CYTAG® Total CGH Labeling Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
Certificates
About this item
Complete kit to digest and label DNA for CGH+SNP arrays.
- Contains CYTAG™ CGH Labeling kit, restriction enzymes AluI and RsaI, and PCR & Gel Clean-up Columns
- Compatible with CGH and CGH+SNP arrays
- Ability to perform total genomic DNA analysis without amplification or complexity reduction
- Provides excellent dynamic analytical range for challenging and complex, heterogeneous samples
- QC benchmarked and validated using high resolution arrays
- Superior DLR scores (0.09-0.12), exceeding industry standards
Comparative genomic hybridization (aCGH) is a powerful diagnostic tool for detecting DNA copy number gains and losses associated with chromosome abnormalities. aCGH provides an understanding of genetic disorders, cancers and other genomic aberrations. With optimized, proprietary reagents, Enzo's CYTAG™ CGH Labeling Kits produce high quality data using as little as 0.25 µg of genomic DNA, without a need for pre-amplification. Bulk quantity kit, 2 x 1000 reactions, is available upon special request.
Ordering information: Contains -0010 size:ENZ-42671-K010: Primers/Reaction Buffer, 1 x 400 µL; Cyanine 3-dUTP Nucleotide Mix, 2 x 50 µL; Cyanine 5-dUTP Nucleotide Mix, 2 x 50 µL; Klenow Exo-DNA Polymerase, 1 x 20 µL; Stop Buffer, 1 x 100 µL; Nuclease-free Water, 1 x 1 mL ENZ-GEN100-0020: PCR & Gel Clean-up columns, 20 testsENZ-GEN108-1000: AluI, 1000 units, 10X Cutting BufferENZ-GEN109-1000: RsaI, 1000 units, 10X Cutting Buffer-0100 size:ENZ-42671-K100: Primers/Reaction Buffer, 1 x 4 mL; Cyanine 3-dUTP Nucleotide Mix, 1 x 1 mL; Cyanine 5-dUTP Nucleotide Mix, 1 x 1 mL; Klenow Exo-DNA Polymerase, 1 x 0.2 mL; Stop Buffer, 1 x 1 mL; Nuclease-free Water, 1 x 10 mL ENZ-GEN100-0200: PCR & Gel Clean-up columns, 200 testsENZ-GEN108-1000: AluI, 1000 units, 10X Cutting BufferENZ-GEN109-1000: RsaI, 1000 units, 10X Cutting Buffer. Also available in custom high throughput formats.