Order Entry
United States
ContactUsLinkComponent
FastDNA™ Kit MP Biomedicals
FastDNA™ Kit MP Biomedicals
Catalog # 75784-604
Supplier:  MP Biomedicals
CAS Number:  
undefined
FastDNA™ Kit MP Biomedicals
Catalog # 75784-604
Supplier:  MP Biomedicals
Supplier Number:  116540400
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Method / Format:
  • Refine target molecule:
    DNA
  • Sample type:
    Plant samples
    Bacteria
    Yeast/Fungi
    Cell culture
  • Target molecule:
    DNA
  • Description:
    FastDNA™ Kit
  • Size:
    100 Preps
  • Environmentally Preferable:
  • Storage Conditions:
    All FastDNA® Kit reagents are shipped ambiently. FastDNA® Kit reagents are stable at room temperature. (15-30 °C)
  • Instrument Compatibility:
    FastPrep-24; FastPrep-96 [Adaptateur QuickPrep (24x2 mL); HiPrep (48x2 mL); CoolPrep (24x2 mL); QuickFlex (96x2 mL); ConeFlex Legacy (allows all FP24 adapters on FP96)]
  • Cat. No.:
    75784-604
  • Supplier no.:
    116540400

Specifications

About this item

Isolation of genomic DNA from plants, animals, bacteria, yeast, algae, and fungi.

  • Rapid and reproducible sample lysis with the FastPrep®-24 or FastPrep® FP120 Instrument
  • Lyse and isolate DNA in less than 30 minutes from a variety of sample types
  • Flexible format eliminates need for separate DNA isolation kits
  • No hazardous organic reagents required

Trusted for over 7 years, the FastDNATM Kit is used with the FastPrep™-24 or FastPrep™FP120 Instrument to lyse and subsequently isolate DNA from up to 200 mg of almost any sample in less than 30 minutes. Lysing Matrix A and three different chaotropic buffers are provided for the homogenization of a wide variety of sample types. The released DNA is purified by a silica based GeneClean™ procedure and is suitable for PCR analysis and other downstream applications.

Components: Lysing Matrix A 100 2 mL tubes, 1/4 Ceramic Spheres 100 spheres, Binding Matrix 66 mL, Concentrated SEWS-M 12 mL, DES 25 mL, CLS VF 90 mL, PPS 25 mL, CLS-TC 110 mL, CLS-Y 110 mL
Material of construction: Matrix A, garnet matrix and one 1/4 inch ceramic sphere
Materials required (not supplied): FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge that can freely spin 2.0 mLtubes, Microcentrifuge tubes (2.0mL and 1.5mL), Rotator or low-speed vortex, (Optional) SPIN Filters and Catch Tubes, 100 (Cat # 2080-800)
Tube compatibility: 2 mL