Order Entry
Puerto Rico
ContactUsLinkComponent
 
Polypropylene Beakers Triple Pour-out
Catalog # 470204-434
undefined
Polypropylene Beakers Triple Pour-out
Catalog # 470204-434
Supplier Number:  25384-154

Specifications

  • Capacity:
    250 ml
  • Cat. No.:
    470204-434
  • Supplier no.:
    25384-154

Specifications

About this item

Transparent Polypropylene Beakers.

  • Three pouring lips for dripless dispensing
  • Graduated ±5% accuracy

Resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents.