3M™ 332 Impact Series Safety Goggles, 3M
55005-305
: 3M
- Description:332 Impact series safety goggles with adjustable headband
- Cat. No.:55005-305
- Lens coating:Uncoated
- Lens tint:Clear
- Supplier no.:40650-00000-10
3M™ 332 series safety impact goggles are a dependable and economical choice that provides a sturdy polycarbonate lens, adjustable elastic strap, and a wide opening to fit over prescription eyewear.
- Economical goggle
- Direct venting system that circulates air and helps minimize fogging
- Over-the-glass coverage
Goggles feature a strap attachment system. These goggles do not offer splash protection.
They are built with an indirect venting system that circulates air and helps minimize fogging, allowing for use in varied environments and conditions.
: Meets ANSI Z87.1-2010.