Order Entry
Puerto Rico
ContactUsLinkComponent
 
Tween® 80 (Polysorbate)
Catalog # 89408-142
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Tween® 80 (Polysorbate)
Catalog # 89408-142
Supplier:  Hardy Diagnostics
Supplier Number:  U180
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    PP bottle
  • Size:
    500 ml
  • Cat. No.:
    89408-142
  • Supplier no.:
    U180

Specifications

About this item

 :
Storage Temperature:Freezer
CAS Number:9005-65-6
MDL Number:MFCD00082107