Tryptone Broth 2%, Hardy Diagnostics
Catalog # 10015-568
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:PC tube
- Description:Tryptone Broth 2%
- Size:13 x 100 mm
- Cat. No.:10015-568
- Volume:2 ml
- Supplier no.:R40
Specifications
About this item
For the performance of the Indole test.
For the performance of the Indole test. Use with Kovacs Reagent.