SterEM™ Tryptic Soy Agar (TSA), USP, with Lecithin and Tween® 80
Catalog # 76200-292
Supplier: Hardy Diagnostics
CAS Number:
Specifications
- Description:SterEM™ Tryptic Soy Agar (TSA) with Lecithin and Tween® 80, USP, irradiated, triple bagged
- Dimensions:15×100 mm Plate
- Cat. No.:76200-292
- pH:7.3 ±0.2 at 25 °C
- Fill volume:26 ml
- Storage temperature:15...30 °C
- Application:Compounding pharmacy, pharmaceutical manufacturing
Specifications
About this item
Sterile plates for environmental monitoring in a clean room or clean bench setting.
- New moisture control packaging format prevents moisture buildup and ensures safe and sterile transfer
- Optically clear polystyrene for distortion free viewing
- Heavy-Duty - Reduces breakage during shipping
- Polystyrene petri plates feature raised rings for stacking ability and prevents sliding
- Sterile by irradiation - validated for sterility assurance level (SAL) of 10⁻⁵
- Triple bagged, two nylon inner bags and one outer Mylar(R) bag that is heat sealed
- Outer bag is VHP impermeable
Tryptic Soy Agar (TSA) with Lecithin and Tween®, USP is recommended for use as a general growth medium for establishing microbiological trends, alerts and action levels in biologically controlled environments. The plates are triple bagged and sterilized by irradiation to promote a higher sterility assurance level.
Certifications: Certified to ISO 13485 - Manufactured in a licensed cGMP facility with the quality management system certified to ISO 13485.
Ordering information: Minimum order of 16 packs.