ContactUsLinkComponent
Tryptic Soy Agar (TSA) with Cycloheximide, Hardy Diagnostics
Tryptic Soy Agar (TSA) with Cycloheximide, Hardy Diagnostics
Catalog # 10146-968
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Tryptic Soy Agar (TSA) with Cycloheximide, Hardy Diagnostics
Catalog # 10146-968
Supplier:  Hardy Diagnostics
Supplier Number:  G70
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Tryptic Soy Agar (TSA) with Cycloheximide
  • Size:
    15 x 100 mm
  • Environmentally Preferable:
  • Minimum Order Quantity:
    10
  • Cat. No.:
    10146-968
  • Supplier no.:
    G70
Specifications
About this item
for the isolation and cultivation of microorganisms while inhibiting Saprophytic fungal organisms.
Hardy Diagnostics Tryptic Soy Agar with Cycloheximide is recommended for use as a general growth for the isolation and cultivation of microorganisms while inhibiting Saprophytic fungal organisms.
Ordering information: Minimum Order of 15 packs