Tetrathionate Broth, Hardy Diagnostics
Catalog # 89407-970
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:PC bottle
- Description:Tetrathionate Broth
- Size:1 L
- Storage Temperature:–2-30°C
- Cat. No.:89407-970
- Volume:1 L
- Supplier no.:U165
Specifications
About this item
For selective enrichment of Salmonella spp.
For selective enrichment of Salmonella spp. Use with iodine-iodide solution.