Tergitol 7 Agar with TTC, Hardy Diagnostics
Catalog # 89501-508
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Plate
- Description:Tergitol 7 Agar with TTC
- Size:15 x 100 mm
- Cat. No.:89501-508
- Supplier no.:G58
Specifications
About this item
For the isolation and differentiation of gram negative bacilli.
For the isolation and differentiation of gram negative bacilli.