ContactUsLinkComponent
TB Base Digestant, Hardy Diagnostics
  10834-146
Product available on GSA Advantage®
 :  
undefined
TB Base Digestant, Hardy Diagnostics
  10834-146
 :  X45
Product available on GSA Advantage®
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Pack type:
    Bottle
  • Description:
    TB Base Digestant
  • Cat. No.:
    10834-146
  • Volume:
    50 ml
  • Supplier no.:
    X45
TB Base Digestant, for the digestion and decontamination of sputum specimens for TB culture
  • For the processing of specimens for AFB culture.
  • Add NALC upon use
For the processing of specimens for AFB culture
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
Add NALC upon use.