TAT Broth
89407-952
Product available on GSA Advantage®
:
- Pack type:PP bottle, 1 L
- Description:TAT Broth
- Size:10/pack
- Storage Temperature:–2...+30 °C
- Minimum Order Quantity:10
- Cat. No.:89407-952
- Volume:490 ml
- Supplier no.:U318
For the detection of microorganisms.
For the detection of microorganisms from topical drugs, cosmetics, and other viscous materials.
: Minimum order of 10 packs.