SterEM™ SabDex (Sabouraud Dextrose) Agar, Irradiated
Catalog # 76200-288
Supplier: Hardy Diagnostics
CAS Number:
Specifications
- Description:SterEM™ Sabdex (Sabouraud Dextrose Agar) agar, USP, irradiated, triple bagged
- Dimensions:15×100 mm (Plate)
- Cat. No.:76200-288
- pH:5.6 ±0.2 at 25 °C
- Fill volume:26 ml
- Storage temperature:15...30 °C
- Application:Compounding pharmacy, pharmaceutical manufacturing
Specifications
About this item
Sterile plates for environmental monitoring.
- New moisture control packaging format
- Prevents moisture buildup and ensures safe and sterile transfer
- Optically clear polystyrene for distortion free viewing
- Heavy-Duty - Reduces breakage during shipping.
- Polystyrene petri plates feature raised rings for stacking ability and prevents sliding
- Sterile by irradiation, validated for Sterility Assurance Level (SAL) of 10⁻⁵
- Triple bagged two nylon inner bags and one outer Mylar® bag that is heat sealed
- Outer bag is VHP impermeable
Sabouraud Dextrose Agar is recommended for the isolation, cultivation, and maintenance of non-pathogenic and pathogenic species of fungi and yeasts, and meet the harmonized USP/EP/JP standards for the microbial examination of non sterile products. The plates are triple bagged and sterilized by irradiation to promote a higher sterility assurance level.
Certifications: Certified to ISO 13485. Manufactured in a licensed cGMP facility with the quality management system certified to ISO 13485.