ContactUsLinkComponent
Regan-Lowe Agar without Cephalexin, Hardy Diagnostics
Regan-Lowe Agar without Cephalexin, Hardy Diagnostics
Catalog # 75802-938
Product available on GSA Advantage®
CAS Number:  
undefined
Regan-Lowe Agar without Cephalexin, Hardy Diagnostics
Catalog # 75802-938
Supplier Number:  A63
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Pack type:
    Plate
  • Description:
    Regan-Lowe Agar without Cephalexin
  • Size:
    15 x 100 mm
  • Cat. No.:
    75802-938
  • Supplier no.:
    A63
Specifications
About this item
For the selective isolation of Bordetella pertussis.
(Charcoal Blood Agar with 10% Horse Blood and w/o Cephalexin) For the selective isolation of Bordetella pertussis. Bordatella are respiratory pathogens, they reside on the mucous membranes of the respiratory tract.