Order Entry
Puerto Rico
ContactUsLinkComponent
CRITERION™ Rappaport-Vassiliadis R10 Broth, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
CRITERION™ Rappaport-Vassiliadis R10 Broth, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
Catalog # 89501-484
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
CRITERION™ Rappaport-Vassiliadis R10 Broth, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
Catalog # 89501-484
Supplier:  Hardy Diagnostics
Supplier Number:  C6730
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Zip-pouch
  • Description:
    Rappaport-Vassiliadis R10 Broth, CRITERION™ Dehydrated Culture Media
  • Cat. No.:
    89501-484
  • Volume:
    2 L
  • Supplier no.:
    C6730

Specifications

About this item

For use in the selective enrichment and isolation of Salmonella spp

  • Wide mouth opening - reduces dust formation by allowing the easy entry of a scoop
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers - for efficient storage
  • Increased solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
  • Complete technical/instruction sheets - available in HUGO software
  • Unparalleled performance - tested for optimal growth

Hardy Diagnostics CRITERION™ Rappaport-Vassiliadis R10 Broth is primarily for use in the selective enrichment and isolation of Salmonella spp. from food and environmental specimens.
CRITERION™ Dehydrated Culture Media by Hardy Diagnostics is formulated to meet or exceed the highest standards.

from food and environmental specimens.

Packaging: Container sizes include: - 2L mylar® bag – pre-measured for a 2 liter batch, no weighing needed!
- 500gm bottle - wide mouth, easy grip bottle, comes with a free scoop!
- 2kg and 10kg bucket - with convenient screw lid