R2A Agar, Hardy Diagnostics
Catalog # 10146-856
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:R2A Agar
- Cat. No.:10146-856
- Volume:400 ml
- Supplier no.:U354
Specifications
About this item
For the detection of heterotrophic bacteria in water, by membrane filtration technique.
- Membrane filtration technique
For the detection of heterotrophic bacteria in water, by membrane filtration technique.