ContactUsLinkComponent
HardyDisks™ AST Novobiocin 5 µg, NB-5, Hardy Diagnostics
HardyDisks™ AST Novobiocin 5 µg, NB-5, Hardy Diagnostics
Catalog # 10789-452
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
HardyDisks™ AST Novobiocin 5 µg, NB-5, Hardy Diagnostics
Catalog # 10789-452
Supplier:  Hardy Diagnostics
Supplier Number:  Z7291
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Description:
    Novobiocin 5ug, NB-5
  • Size:
    1 x 50 disks
  • Cat. No.:
    10789-452
  • Supplier no.:
    Z7291
Specifications
About this item
Used to identify Staphylococcus saprophyticus.
Used to identify Staphylococcus saprophyticus, which is resistant to Novobiocin 5ug (zones less than or equal to 16mm)
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing from the single metal disk dispense.
Delivery: Freeze on arrival