ContactUsLinkComponent
SCI Needle, Nichrome, with Aluminum Handle
SCI Needle, Nichrome, with Aluminum Handle
Catalog # 10063-754
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
undefined
SCI Needle, Nichrome, with Aluminum Handle
Catalog # 10063-754
Supplier:  Hardy Diagnostics
Supplier Number:  3060
Product available on GSA Advantage®
Specifications
  • Description:
    Needle, nichrome, with aluminum handle with insulated sleeve
  • Size:
    Pack of 5
  • Cat. No.:
    10063-754
  • Supplier no.:
    3060
Specifications
About this item
Used to streak various liquids, suspensions, bacteria, and other specimen on to culture media and/or petri dishes for purposes of study and growth.