ContactUsLinkComponent
CRITERION™ Muller-Kauffmann Tetrathionate Broth, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
CRITERION™ Muller-Kauffmann Tetrathionate Broth, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
  89501-494
Product available on GSA Advantage®
 :  
undefined
CRITERION™ Muller-Kauffmann Tetrathionate Broth, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
  89501-494
 :  C8481
Product available on GSA Advantage®
 :  
  • Pack type:
    Bottle, wide mouth
  • Description:
    Muller-Kauffmann Tetrathionate Broth
  • Size:
    500 gm
  • Minimum Order Quantity:
    2
  • Cat. No.:
    89501-494
  • Supplier no.:
    C8481
recommended for the isolation of Salmonella and the suppression of Proteus spp.
  • Wide mouth opening - reduces dust formation by allowing the easy entry of a scoop
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers - for efficient storage
  • Increased solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
  • Competitive pricing - call for a price quote for large volume purchases!
  • Complete technical/instruction sheets - available in HUGO software
  • Unparalleled performance - tested for optimal growth
Hardy Diagnostics CRITERION™ Muller-Kauffmann Tetrathionate Broth is recommended for the isolation of Salmonella and the suppression of Proteus spp.
CRITERION™ Dehydrated Culture Media by Hardy Diagnostics is formulated to meet or exceed the highest standards.
Container sizes include: 2L mylar® bag – pre-measured for a 2 liter batch, no weighing needed; 500gm bottle - wide mouth, easy grip bottle, comes with a free scoop; 2kg and 10kg bucket - with convenient screw lid
Also see our selection of petri plates, glass screw-cap tubes, animal blood, media supplements and antibiotics.
 : Minimum order of two.