Modified Buffered Peptone Water, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
Catalog # 10033-084
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:Modified Buffered Peptone Water Dehydrated Culture Media
- Size:2 kg
- Cat. No.:10033-084
- Supplier no.:C6352
Specifications
About this item
Pre-enrichment for increased recovery of Salmonella species in foods.
- Wide mouth opening - reduces dust formation by allowing the easy entry of a scoop
- Convenient handgrip - 'no slip' finger holds
- Stackable containers - for efficient storage
- Increased solubility - dissolves quickly
- Custom and bulk products available - to your specifications
- Competitive pricing - call for a price quote for large volume purchases!
- Complete technical/instruction sheets - available in HUGO software
- Unparalleled performance - tested for optimal growth
Hardy Diagnostics CRITERION™ Modified Buffered Peptone Water is used as a pre-enrichment for increased recovery of Salmonella species in foods.
CRITERION™ Dehydrated Culture Media by Hardy Diagnostics is formulated to meet or exceed the highest standards.
Container sizes include: - 2L mylar® bag – pre-measured for a 2 liter batch, no weighing needed!
- 500gm bottle - wide mouth, easy grip bottle, comes with a free scoop!
- 2kg and 10kg bucket - with convenient screw lid
Also see our selection of petri plates, glass screw-cap tubes, animal blood, media supplements and antibiotics.