MIL (Motility, Indole, Lysine deaminase, and Decarboxylase) Medium, Hardy Diagnostics
Catalog # 10053-814
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Tube
- Description:MIL Medium, HDx, 16x100mm tube
- Size:16 x 100 mm
- Cat. No.:10053-814
- Volume:8 ml
- Supplier no.:Q09
Specifications
About this item
For the determination of motility, indole, lysine deamination, and lysine decarboxylation of gram negative bacilli.
For the determination of motility, indole, lysine deamination, and lysine decarboxylation of gram negative bacilli.