Middlebrook 7H11, Hardy Diagnostics
10158-950
Product available on GSA Advantage®
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:Middlebrook 7H11, deep fill
- Size:15×100 mm
- Cat. No.:10158-950
- Supplier no.:W35
For the cultivation of Mycobacterium spp.
- Optically clear polystyrene for distortion free viewing
- Heavy-Duty - Reduces breakage during shipping
- Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
- Packaged in sleeves of 10 plates in a "breathable" bag that prevents buildup of condensation and excess moisture
Middlebrook 7H11 agar is recommended for use in the isolation and cultivation of Mycobacterium species.