LJ (Lowenstein Jensen) Gruft Slant, Hardy Diagnostics
Catalog # 10789-714
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Tube
- Description:LJ Gruft Slant, HDx
- Size:20 x 125 mm
- Cat. No.:10789-714
- Volume:10 ml
- Supplier no.:C23
Specifications
About this item
For the selective isolation of mycobacteria.
For the selective isolation of mycobacteria. Contains penicillin, nalidixic acid, and RNA.