ContactUsLinkComponent
Listeria Enrichment Broth, Modified, Hardy Diagnostics
  89407-744
 :  
undefined
Listeria Enrichment Broth, Modified, Hardy Diagnostics
  89407-744
 :  U167
 :  
  • Pack type:
    PC bottle
  • Description:
    Listeria Enrichment Broth, Modified
  • Size:
    225 mL
  • Storage Temperature:
    2-8°C
  • Cat. No.:
    89407-744
  • Volume:
    225 ml
  • Supplier no.:
    U167
For the selective isolation of Listeria spp.
For the selective isolation of Listeria spp.