$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Lugol's iodine solution
Lugol's iodine solution, Dobell and O'Connor stain, used in parasitology
Lugol's iodine solution, Dobell and O'Connor stain, used in parasitology
10790-370
:
Hardy Diagnostics
Product available on GSA Advantage®
:
Lugol's iodine solution, Dobell and O'Connor stain, used in parasitology
10790-370
:
Hardy Diagnostics
:
Z66
Product available on GSA Advantage®
:
Pack type:
Glass bottle, amber
Size:
15 ml
Cat. No.:
10790-370
Supplier no.:
Z66
:
:
GHS07