Frey Medium Agar, Hardy Diagnostics
Catalog # 10124-542
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:Frey Medium Agar, HDx
- Size:15 x 60 mm
- Minimum Order Quantity:10
- Cat. No.:10124-542
- Volume:11 ml
- Supplier no.:G381
Specifications
About this item
Recommended for use in the cultivation and isolation of Mycoplasma species.
Recommended for use in the cultivation and isolation of Mycoplasma species.
Ordering information: Minimum order of 10 packs