HardyDisks™ AST Erythromycin, E-15, Hardy Diagnostics
Supplier: Hardy Diagnostics
Certificates
About this item
Erythromycin, E-15, HardyDisks™ AST, for susceptibility testing, freeze on arrival
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing. The HardyDisk™ AST cartridges are designed to be fully compatible with the BBL single or multiple disk dispensers.
Delivery: Freeze on arrival.