EnviroBootie™, Hardy Diagnostics
Supplier: Hardy Diagnostics
Certificates
About this item
The next generation sample collection device for the monitoring of Salmonella and environmental contamination.
EnviroBootie™ is the next generation sample collection device for the monitoring of Salmonella and environmental contamination in flock floor litter and other surfaces.
EnviroBootie™ is hydrated with double strength skim milk, and packaged in a polyethylene bag.
Frequently Bought Together