Order Entry
Puerto Rico
ContactUsLinkComponent
CompactDry™ LS, Listeria spp, Hardy Diagnostics
CompactDry™ LS, Listeria spp, Hardy Diagnostics
Catalog # 10789-408
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
CompactDry™ LS, Listeria spp, Hardy Diagnostics
Catalog # 10789-408
Supplier:  Hardy Diagnostics
Supplier Number:  LS100
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Description:
    CompactDry LS, HDx
  • Size:
    25 packs of 4 plates
  • Storage Conditions:
    15…25 °C
  • Cat. No.:
    10789-408
  • Supplier no.:
    LS100

Specifications

About this item

For the isolation, presumptive identification and enumeration of Listeria spp.

  • Room temperature storage
  • Self diffusing media
  • Stackable
  • Write-on area
  • Easy to handle and pick up
  • Simple to pick colonies for subculturing
  • Easy read-out of results

CompactDry™ LS for the isolation, presumptive identification and enumeration of Listeria spp

This product is not intended to be used for the diagnosis of human disease.

Colonies of Listeria will turn blue due to the presence of chromogenic substances.