Chocolate Agar with Bacitracin, Hardy Diagnostics
Catalog # 10053-844
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Plate
- Description:Chocolate agar w/Bacitracin
- Size:15×100 mm
- Cat. No.:10053-844
- Volume:1 L
- Supplier no.:E11
Specifications
About this item
Chocolate Agar with Bacitracin for Haemophilus isolation.
For selective isolation of Haemophilus spp. from respiratory specimens. Inhibits most staphylococci, streptococci, neisseria, and other normal flora.
Ordering information: Order by the pack of 10