Hydrogen peroxide solution 15%
15 ml
Supplier: HARDY DIAGNOSTICS CA
Supplier #: Z262
Hydrogen peroxide solution 15%
Catalog # 10063-756
Supplier: HARDY DIAGNOSTICS CA
CAS Number: 7722-84-1
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
A high strength 15% solution of hydrogen peroxide especially recommended for the detection of catalase activity in anaerobic bacteria.
- CAS Number:7722-84-1
- Formula:H₂O₂
- MDL Number:MFCD00011333
- Molecular Weight:34.01
- :Density: 1.11 g/cm³ (20 °C)
Documents
Specifications
Specifications
- Size:15 ml



