Order Entry
Puerto Rico
ContactUsLinkComponent
Buffer No. 16, Hardy Diagnostics
Buffer No. 16, Hardy Diagnostics
Catalog # 10693-110
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Buffer No. 16, Hardy Diagnostics
Catalog # 10693-110
Supplier:  Hardy Diagnostics
Supplier Number:  U558
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    PC bottle
  • Description:
    Buffer No. 16, USP
  • Cat. No.:
    10693-110
  • Volume:
    500 ml
  • Supplier no.:
    U558

Specifications

About this item

For the use in determination of antimicrobial activity.

For the use in determination of antimicrobial activity. Formulated according to USP <81>.
MINIMUM ORDER: 5 packs