Order Entry
Puerto Rico
ContactUsLinkComponent
 
Brain Heart Infusion Broth (BHI) with 15% Glycerol, Hardy Diagnostics
  10119-248
Product available on GSA Advantage®
 :  
undefined
Brain Heart Infusion Broth (BHI) with 15% Glycerol, Hardy Diagnostics
  10119-248
 :  D114
Product available on GSA Advantage®
 :  

 

  • Pack type:
    Vial
  • Description:
    BHI Broth w/15% Glycerol
  • Size:
    15 x 45 mm
  • Cat. No.:
    10119-248
  • Volume:
    2 ml
  • Supplier no.:
    D114

 

 

A cryopreservative for the freezing of microorganisms.

A cryopreservative for the freezing of microorganisms.