CRITERION™ Barney Miller Broth with Indicator, Hardy Diagnostics
Catalog # 75802-966
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Bottle, wide mouth
- Description:BMB with Indicator, HDx
- Cat. No.:75802-966
- Volume:500 g
- Supplier no.:C8991
Specifications
About this item
Barney Miller Broth with Indicator for the isolation and detection of beer spoilage microorganisms
- Wide mouth opening - reduces dust formation by allowing the easy entry of a scoop
- Convenient handgrip - 'no slip' finger holds
- Stackable containers - for efficient storage
- Increased solubility - dissolves quickly
- Custom and bulk products available - to your specifications
- Competitive pricing - call for a price quote for large volume purchases!
- Complete technical/instruction sheets - available in HUGO software
- Unparalleled performance - tested for optimal growth
Hardy Diagnostics CRITERION™ Barney Miller Broth with Indicator is recommended for the isolation and detection of beer spoilage microorganisms, specifically lactic acid bacteria
CRITERION™ Dehydrated Culture Media by Hardy Diagnostics is formulated to meet or exceed the highest standards.
Container sizes include: - 2L mylar® bag – pre-measured for a 2 liter batch, no weighing needed!
- 500gm bottle - wide mouth, easy grip bottle, comes with a free scoop!
- 2kg and 10kg bucket - with convenient screw lid
Also see our selection of petri plates, glass screw-cap tubes, animal blood, media supplements and antibiotics.