Order Entry
Puerto Rico
ContactUsLinkComponent
CRITERION™ Barney Miller Agar with Indicator, Hardy Diagnostics
CRITERION™ Barney Miller Agar with Indicator, Hardy Diagnostics
Catalog # 75802-964
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
CRITERION™ Barney Miller Agar with Indicator, Hardy Diagnostics
Catalog # 75802-964
Supplier:  Hardy Diagnostics
Supplier Number:  C9201
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Bottle, wide mouth
  • Description:
    BMA with Indicator Base, HDx
  • Cat. No.:
    75802-964
  • Volume:
    500 g
  • Supplier no.:
    C9201

Specifications

About this item

Barney Miller Agar with Indicator for the detection and enumeration of beer spoilage microorganisms.

  • Wide mouth opening - reduces dust formation by allowing the easy entry of a scoop
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers - for efficient storage
  • Increased solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
  • Competitive pricing - call for a price quote for large volume purchases!
  • Complete technical/instruction sheets - available in HUGO software
  • Unparalleled performance - tested for optimal growth

Hardy Diagnostics CRITERION™ Barney Miller Agar with Indicator is recommended for the detection and enumeration of beer spoilage microorganisms.

CRITERION™ Dehydrated Culture Media by Hardy Diagnostics is formulated to meet or exceed the highest standards.

Container sizes include: - 2L mylar® bag – pre-measured for a 2 liter batch, no weighing needed!
- 500gm bottle - wide mouth, easy grip bottle, comes with a free scoop!
- 2kg and 10kg bucket - with convenient screw lid