Order Entry
Puerto Rico
ContactUsLinkComponent
Hygiena ALLER-Snap™ High-Sensitivity Protein Residue Test, Hardy Diagnostics
Hygiena ALLER-Snap™ High-Sensitivity Protein Residue Test, Hardy Diagnostics
Catalog # 10062-760
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Hygiena ALLER-Snap™ High-Sensitivity Protein Residue Test, Hardy Diagnostics
Catalog # 10062-760
Supplier:  Hardy Diagnostics
Supplier Number:  ALS100
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Description:
    ALLER-Snap, Protein Test
  • Size:
    100 per box
  • Cat. No.:
    10062-760
  • Supplier no.:
    ALS100

Specifications

About this item

ALLER-Snap™ detects protein residues left on a surface after cleaning.

  • All-in-one test device
  • Built-in color chart
  • Pre-wetted swab
  • Sensitive liquid-stable reagent
  • Generate visible color results in 15-30 minutes
  • Small simple design
  • 18 month shelf-life at room or refrigerated temperature

ALLER-Snap™ is a quick and easy way to accurately monitor the cleanliness of surfaces to help ensure product quality

Simply swab a surface, release the reagent and if protein residue is present the reagent will turn purple. The color change provides a semi-quantitative measure of the surface cleanliness. The more contamination present, the quicker the color change to purple and the darker the color. ALLER-Snap™ quickly validates the hygiene of a surface, allowing immediate corrective action to be taken if necessary.

Sensitivity:
Detects down to 3 µg of protein residue
Enhanced sensitivity by incubating at 55°C for 15 minutes or 30 minutes at 37°C

Benefits:
Ready-to-use and Convenient
Simple, easy-to-use and interpret
Consistent sample collection
Maximum sample recovery
Breaks through biofilms
No separate wetting agent required
Suitable for allergen monitoring
Store at room temperature
Enables immediate corrective action thus preventing waste and non-compliance issues
Provides evidence for due diligence and compliance
Effective for trainings and promotes best practices
Required instrumentation: Incubator
Small size
Recyclable plastic
Low carbon footprint