ContactUsLinkComponent
Organon Teknika Airway Needle
Organon Teknika Airway Needle
Catalog # 10153-952
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
undefined
Organon Teknika Airway Needle
Catalog # 10153-952
Supplier:  Hardy Diagnostics
Supplier Number:  33766
Product available on GSA Advantage®
Specifications
  • Sterility:
    Sterile
  • Description:
    Airway needle for use with blood culture bottles
  • Size:
    100 per box
  • Cat. No.:
    10153-952
  • Supplier no.:
    33766
Specifications
About this item
Sterile subculturing disposable for use with blood culture bottles.
  • Easy to use
  • Disposable
To aerate with filtered air and to prevent build up of gas pressure during incubation.
Applications: Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, compounding pharmacy, pharmaceutical manufacturing