Specifications
- Size:1600 units
- Enzyme Name:Thermostable FEN1
- Cat. No.:103218-938
- Supplier no.:M0645S
Specifications
About this item
Thermostable Flap Endonuclease 1, FEN1, catalyzes the cleavage of 5' DNA flaps from branched double stranded DNA substrates, creating a 5' phosphate terminus
- Thermostable DNA and RNA endonuclease
- Catalyzes the cleavage of 5' DNA and RNA flaps from branched double stranded DNA substrates
- FEN 1 products can be ligated by DNA ligase to create double stranded DNA
- Heat inactivation not possible
FEN1 products can be ligated by DNA ligase to create double stranded DNA. In vivo, FEN1 is an essential component of the Okazaki fragment maturation pathway, and also plays a role in base excision repair.