Order Entry
Puerto Rico
ContactUsLinkComponent
2-Propanol, anhydrous, Surgipath® for histology and pathology
Flammable
Irritant
2-Propanol, anhydrous, Surgipath® for histology and pathology
Catalog # 75810-350
Supplier:  Leica Biosystems
CAS Number:  
undefined
Flammable
Irritant
2-Propanol, anhydrous, Surgipath® for histology and pathology
Catalog # 75810-350
Supplier:  Leica Biosystems
Supplier Number:  3803660
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    4 gal.
  • Cat. No.:
    75810-350
  • Supplier no.:
    3803660

Specifications

About this item

 :
 :60.1 g/mol
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
 :GHS02
 :GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674