

Reagent alcohol ≥95%, Surgipath® ISO for histology and pathology
Catalog # 10015-097
Supplier: Leica Biosystems
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:1 gal.
- Cat. No.:10015-097
- Supplier no.:3803682
Specifications
About this item
:
Flash Pt:55 °F
UN:1987
ADR:3,II
:GHS02
:GHS08
CAS Number:64-17-5
MDL Number:MFCD00003568