ContactUsLinkComponent
Surgipath® Snowcoat® Beveled-Edge Microscope Slides, Leica
Surgipath® Snowcoat® Beveled-Edge Microscope Slides, Leica
Catalog # 75809-504
Supplier:  Leica Biosystems
undefined
Surgipath® Snowcoat® Beveled-Edge Microscope Slides, Leica
Catalog # 75809-504
Supplier:  Leica Biosystems
Supplier Number:  3800373
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Snowcoat® clipped corner slides, colored ends
  • Cat. No.:
    75809-504
  • Supplier no.:
    3800373
Specifications
About this item
Snowcoat Beveled Edge Slides feature beveled edges and corners that make for easier handling.
  • Beveled edges
  • Easy handling
  • For blood smears
Slides are good for blood smears, producing a feathered edge. Painted white surface facilitates quality marking adherence. Each slide is washed and carefully inspected to ensure quality.