Pullulan (from Aureobasidium pullulans), white powder
Catalog # IC15394401
Supplier: MP Biomedicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:1 g
- Cat. No.:IC15394401
- Supplier no.:15394401
Specifications
Specification Test Results
- Loss on drying:0,032
- Ash:0,0018
- Viscosity (10% w/w aq. soln @ 30°):124 mm^2/s
- Lead:<1 mg/kg
- Arsenic:<0.5 mg/kg
- Total plate count:1000/g
- Yeast & Molds:1000/g
- Coliforms:Absence/25g
- Salmonella:Negative/25g
- Solubility (5% DI Water):Clear colorless solution
- Assay (dry basis):96% (of glucan)
- pH:5.3 (10% soln)
Specification Test Results
About this item
CAS Number:9057-02-7
:
MDL Number:MFCD00081940
Storage Temperature:Ambient
Formula:(C₆H₁₂O₅)n