Order Entry
Puerto Rico
ContactUsLinkComponent
Magnesium chloride hexahydrate, Molecular Biology Grade, Calbiochem®
Magnesium chloride hexahydrate, Molecular Biology Grade, Calbiochem®
Catalog # 80503-016
Supplier:  MilliporeSigma
CAS Number:  
Magnesium chloride hexahydrate, Molecular Biology Grade, Calbiochem®
Catalog # 80503-016
Supplier:  MilliporeSigma
Supplier Number:  442611-500GM
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    500 g
  • Cat. No.:
    80503-016
  • Supplier no.:
    442611-500GM

Specifications

Specification Test Results

  • Purity:
    ≥99% by complexometry
  • Contaminants:
    DNases, proteases, RNases: none detected
  • Heavy metals:
    ≤0.0005% (as Pb)

Specification Test Results

About this item

 :
 :203.3 g/mol
Merck Index:13,05684
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:117 °C
Density:1.57 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7791-18-6
Formula:MgCl₂·6H₂O
MDL Number:MFCD00149781