

Succinylcholine chloride dihydrate USP Reference Standard
Catalog # 101096-186
Supplier: USP
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:500 mg
- Cat. No.:101096-186
- Supplier no.:1623502
Specifications
Specification Test Results
- Appearance:White Crystalline Powder
- pH:4
- Solubility:Slightly soluble in ethanol and in chloroform; sparingly soluble in benzene; practically insoluble in ether and in acetone
Specification Test Results
About this item
:
:397.34 g/mol
Melting Pt:156…160 °C
UN:2811
ADR:6.1,III
:GHS06
:GHS08
CAS Number:6101-15-1
Formula:C₁₄H₃₀Cl₂N₂O₄.2H₂O
MDL Number:MFCD00150564