Order Entry
Puerto Rico
ContactUsLinkComponent
Brain Heart Infusion Agar, Life Technologies
Brain Heart Infusion Agar, Life Technologies
Catalog # 90000-066
Supplier:  BD
CAS Number:  
Brain Heart Infusion Agar, Life Technologies
Catalog # 90000-066
Supplier:  BD
Supplier Number:  211065
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Brain Heart Infusion Agar
  • Size:
    500 g
  • Cat. No.:
    90000-066
  • Supplier no.:
    211065

Specifications

About this item

Brain Heart Infusion Agar is a general-purpose culture medium which supports the growth of a wide variety of bacterial and fungal species including many types of pathogens, such as streptococci and pneumococci.

  • Brain heart infusion, peptones, and dextrose provide nutrients
  • Dextrose is the carbohydrate source for fermentation
  • Disodium phosphate acts as a buffering agent

Brain Heart Infusion (BHI) Agar is a general-purpose medium suitable for the cultivation of a wide variety of organism types, including bacteria, yeasts, and molds. With the addition of 10% sheep blood, it is used for the isolation and cultivation of a wide variety of fungal species, including systemic fungi, from clinical and nonclinical sources.