ContactUsLinkComponent
Difco™ Antimicrobic Vial A Chlortetracycline, BD Diagnostics
  90002-632
 :  BD
 :  
Difco™ Antimicrobic Vial A Chlortetracycline, BD Diagnostics
  90002-632
 :  BD
 :  233331
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Difco™ Antimicrobic Vial A Chlortetracycline
  • Size:
    10 mL
  • Environmentally Preferable:
  • Cat. No.:
    90002-632
  • Supplier no.:
    233331
Difco™ Antimicrobic Vial A, containing chlortetracycline, is used in preparing selective microbiological culture media.
  • Contains sterile, desiccated chlortetracycline
  • Applicable for use in various media requiring this antibiotic
  • Preferred in Cooke Rose Bengal Agar due to increased stability
Antimicrobic Vial A was originally used in preparing DTM Agar described by Taplin, Azias, Rebell, and Blank for the isolation of dermatophytes.