Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
BBL™ Agar, Select, BD

BBL™ Agar, Select, BD

BBL™ Agar, Select, 500 g
Catalog # 90005-196
Supplier:   BD
Supplier #:  299340
BBL™ Agar, Select, BD
Catalog # 90005-196
Supplier:  BD
CAS Number:  
BBL™ Agar, Select, BD

BBL™ Agar, Select, 500 g

CAS Number:  
Price
$558.00
Log in for availability
Each (500g)
Minimum order quantity is 1
Product Details & Documents

Agar, Select is used as a solidifying agent for culture media.

  • Selective media for culture cultivation

Agar, Select is carefully monitored for cultural response, solubility, and gelation temperature.

Documents

Specifications

Specifications

  • Description:
    BBL™ Agar, Select
  • Size:
    500 g

Specifications