Thiourea on resin (MP), beads, macroporous 100-200 mesh Loading: 1.7-1.8 mmol/g
Supplier: Supra Sciences
Certificates
About this item
CAS Number:62-56-6
:
Formula:NH₂CSNH₂
MDL Number:MFCD00008067
:76.12 g/mol
Specification Test Results
- Colour:Off-white
- Form:Granule
- Loading:1.7-1.8 mmol/g
- Particle size:100-200 mesh