Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Thiol resin
Thiol resin (SI), beads Avg 40-62 µm Loading: 1.0-1.2 mmol/g
Thiol resin (SI), beads Avg 40-62 µm Loading: 1.0-1.2 mmol/g
:
Supra Sciences
Thiol resin (SI), beads Avg 40-62 µm Loading: 1.0-1.2 mmol/g
:
Supra Sciences
:
Colour:
Off-white
Form:
Granule
Loading:
1.0-1.2 mmol/g
Particle size:
40-62 μm
ghs_SPSI19
Thiol