$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Trisamine resin
Trisamine resin (SI), beads Avg. 40-62 µm Loading: 1.0-1.2 mmol/g
Trisamine resin (SI), beads Avg. 40-62 µm Loading: 1.0-1.2 mmol/g
Trisamine resin (SI), beads Avg. 40-62 µm Loading: 1.0-1.2 mmol/g
Supplier:
Supra Sciences
Certificates
Expand image
Trisamine resin (SI), beads Avg. 40-62 µm Loading: 1.0-1.2 mmol/g
Supplier:
Supra Sciences
About this item
:
Specification Test Results
Colour:
Off-white
Form:
Granule
Loading:
1.0-1.2 mmol/g
Particle size:
40-62 μm
Specification Test Results
Related Information
Documentation
SDS
ghs_SPSI20
Datasheet
Trisamine
Documentation